Gene Transcripts
- Gene Symbol
- KSR1
- Entrez Gene ID
- 8844
- Gene Name
- kinase suppressor of ras 1
- Gene Aliases
- KSR, RSU2
- Chromosome Location
- Chr. 17: 27456441 - 27626438 on Build GRCh38
- UniGene ID
- Hs.133534
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014238.1 |
NP_055053.1
|
8, 9 |
1172 |
|
XM_006722154.3 |
XP_006722217.1
|
7, 8 |
1324 |
|
XM_011525429.2 |
XP_011523731.1
|
7, 8 |
1324 |
|
XM_011525430.2 |
XP_011523732.1
|
7, 8 |
1326 |
|
XM_011525431.2 |
XP_011523733.1
|
7, 8 |
1326 |
|
XM_017025267.1 |
XP_016880756.1
|
7, 8 |
1324 |
|
XM_017025268.1 |
XP_016880757.1
|
7, 8 |
1324 |
|
XM_017025269.1 |
XP_016880758.1
|
7, 8 |
1324 |
|
XM_017025270.1 |
XP_016880759.1
|
6, 7 |
1059 |
|
XM_017025271.1 |
XP_016880760.1
|
8, 9 |
2031 |
|
XM_017025272.1 |
XP_016880761.1
|
8, 9 |
2014 |
|
XM_017025273.1 |
XP_016880762.1
|
7, 8 |
1837 |
|
XM_017025274.1 |
XP_016880763.1
|
7, 8 |
1040 |
|
XM_017025275.1 |
XP_016880764.1
|
7, 8 |
1027 |
|
XM_017025276.1 |
XP_016880765.1
|
8, 9 |
1979 |
|
XM_017025277.1 |
XP_016880766.1
|
3, 4 |
722 |
|
XM_017025278.1 |
XP_016880767.1
|
3, 4 |
732 |
|
XM_017025279.1 |
XP_016880768.1
|
7, 8 |
1341 |
|
XM_017025280.1 |
XP_016880769.1
|
7, 8 |
1341 |
|
GenBank mRNA |
BC035488.1 |
- |
8, 9 |
2150 |
|
BC042106.1 |
- |
8, 9 |
1171 |
|
U43586.1 |
- |
4, 5 |
412 |
|
View Full Product Details