Gene Transcripts

Gene Symbol
TRPC7
Entrez Gene ID
57113
Gene Name
transient receptor potential cation channel subfamily C member 7
Gene Aliases
TRP7
Chromosome Location
Chr. 5: 136213311 - 136365476 on Build GRCh38
UniGene ID
Hs.591263
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167577.1 NP_001161049.1 3 1071
NM_020389.2 NP_065122.1 4 1254
GenBank mRNA AJ272034.1 - 3 1032
AJ421783.1 - 3 1032
AJ549088.1 - 2 849
BC128185.1 - 3 1032 40113370
BC128186.1 - 3 1032

View Full Product Details