Gene Transcripts

Gene Symbol
STYXL1
Entrez Gene ID
51657
Gene Name
serine/threonine/tyrosine interacting-like 1
Gene Aliases
DUSP24, MK-STYX, MKSTYX
Chromosome Location
Chr. 7: 75995820 - 76048709 on Build GRCh38
UniGene ID
Hs.11615
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317785.1 NP_001304714.1
9 1318
NM_001317786.1 NP_001304715.1
9 1168
NM_001317787.1 NP_001304716.1
7 877
NM_001317788.1 NP_001304717.1
7 830
NM_001317789.1 NP_001304718.1
7 827
NM_016086.2 NP_057170.1
9 1315
NR_134486.1
7 906
NR_134487.1
7 860
XM_011516292.2 XP_011514594.1
8 1048
XM_011516293.2 XP_011514595.1
9 1161
XM_011516300.1 XP_011514602.1
8 971
XM_017012296.1 XP_016867785.1
10 1275
XM_017012297.1 XP_016867786.1
8 1051
XM_017012298.1 XP_016867787.1
8 1011
XM_017012299.1 XP_016867788.1
10 1228
XM_017012300.1 XP_016867789.1
10 1195
XM_017012301.1 XP_016867790.1
8 1015
XM_017012302.1 XP_016867791.1
8 1018
GenBank mRNA AF069762.1 -
9 1311
AF188202.1 -
5 549
AF188203.1 -
5 579
AF188204.1 -
6 692
AY927581.1 - LIT3580
AY927581
9 1072
BC024035.2 -
9 1278 2988900
BX537896.1 -
7 857

View Full Product Details