Gene Transcripts

Gene Symbol
USP47
Entrez Gene ID
55031
Gene Name
ubiquitin specific peptidase 47
Gene Aliases
TRFP
Chromosome Location
Chr. 11: 11841423 - 11959325 on Build GRCh38
UniGene ID
Hs.577256
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282659.1 NP_001269588.1 7, 8 1590
NM_017944.3 NP_060414.3 6, 7 1386
XM_005252997.1 XP_005253054.1 8, 9 1650
XM_017017953.1 XP_016873442.1 8, 9 1706
GenBank mRNA AK027283.1 - 498
AK027362.1 - 6, 7 791
AK092290.1 - 8, 9 1007
AK127264.1 - 6, 7 1386
AK309240.1 - 7, 8 1054

View Full Product Details