Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282659.1 | NP_001269588.1 | 16 | 2592 | |
NM_017944.3 | NP_060414.3 | 15 | 2388 | ||
XM_005252997.1 | XP_005253054.1 | 17 | 2652 | ||
XM_011520196.1 | XP_011518498.1 | 9 | 1767 | ||
XM_017017953.1 | XP_016873442.1 | 17 | 2708 | ||
XM_017017954.1 | XP_016873443.1 | 9 | 2915 | ||
GenBank mRNA | AK027362.1 | - | 15 | 1793 | |
AK074076.1 | - | 1 | 80 | ||
AK092290.1 | - | 17 | 2008 | ||
AK127264.1 | - | 16 | 2388 | ||
BG752249.1 | - | 422 |