Gene Transcripts

Gene Symbol
CARD6
Entrez Gene ID
84674
Gene Name
caspase recruitment domain family member 6
Gene Aliases
CINCIN1
Chromosome Location
Chr. 5: 40841190 - 40855354 on Build GRCh38
UniGene ID
Hs.200242
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032587.3 NP_115976.2 3 3239
XM_017009989.1 XP_016865478.1 2 2792
XM_017009990.1 XP_016865479.1 4 3473
GenBank mRNA AF356193.1 - 3 3363

View Full Product Details