Gene Transcripts

Gene Symbol
THOP1
Entrez Gene ID
7064
Gene Name
thimet oligopeptidase 1
Gene Aliases
EP24.15, MEPD_HUMAN, MP78, TOP
Chromosome Location
Chr. 19: 2785460 - 2813604 on Build GRCh38
UniGene ID
Hs.78769
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003249.4 NP_003240.1 3 445
GenBank mRNA BC000135.2 - 3 371
BC002391.2 - 3 397 2819803
BC013878.2 - 3 361 3690133
BU849088.1 - 3 445
U29366.1 - 3 329
Z50115.1 - 3 369

View Full Product Details