Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001848.2 | NP_001839.2 | 4 | 554 | |
GenBank mRNA | AK298968.1 | - | 4 | 533 | |
AK299396.1 | - | 4 | 536 | ||
AK307989.1 | - | 4 | 533 | ||
BC052575.1 | - | 4 | 534 | 6598940 | |
CN369026.1 | - | 4 | 552 | ||
X15879.1 | - | 4 | 500 |