Gene Transcripts

Gene Symbol
CTSB
Entrez Gene ID
1508
Gene Name
cathepsin B
Gene Aliases
APPS, CPSB
Chromosome Location
Chr. 8: 11842524 - 11868150 on Build GRCh38
UniGene ID
Hs.520898
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317237.1 NP_001304166.1 4, 5 483
NM_001908.4 NP_001899.1 3, 4 395
NM_147780.3 NP_680090.1 5, 6 557
NM_147781.3 NP_680091.1 4, 5 514
NM_147782.3 NP_680092.1 4, 5 483
NM_147783.3 NP_680093.1 4, 5 469
XM_006716244.2 XP_006716307.1 5, 6 581
XM_006716245.2 XP_006716308.1 3, 4 685
XM_011543812.2 XP_011542114.1 5, 6 490
XM_017013097.1 XP_016868586.1 4, 5 471
XM_017013098.1 XP_016868587.1 4, 5 471
XM_017013099.1 XP_016868588.1 5, 6 513
XM_017013100.1 XP_016868589.1 4, 5 415
GenBank mRNA AK075393.1 - 3, 4 371
AK092070.1 - 4, 5 406
AK130184.1 - 3, 4 332
AK290239.1 - 3, 4 332
AK296845.1 - 3, 4 334
AL543654.3 - 4, 5 443
BC010240.1 - 4, 5 412 4154307
BC095408.1 - 3, 4 332 30334082
BG770805.1 - 5, 6 466
BM008741.1 - 3, 4 371
BX647765.1 - 4, 5 374
BX647952.1 - 3, 4 335
CB997355.1 - 3, 4 409
DC386022.1 - 3, 4 395
L16510.1 - 4, 5 397
M14221.1 - 4, 5 414

View Full Product Details