Gene Transcripts

Gene Symbol
PPA1
Entrez Gene ID
5464
Gene Name
pyrophosphatase (inorganic) 1
Gene Aliases
HEL-S-66p, IOPPP, PP, PP1, SID6-8061
Chromosome Location
Chr. 10: 70202830 - 70233434 on Build GRCh38
UniGene ID
Hs.437403
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021129.3 NP_066952.1 3 263
GenBank mRNA AB026723.1 - 3 186
AF092439.1 - 3 233
AF108211.1 - 3 144
AF119665.1 - 3 233
AF154065.1 - 3 201
AF217186.1 - 3 156
AK058193.1 - 3 256
BC001022.2 - 3 238 3345496
BC061581.1 - 3 246 6526628
BC105034.1 - 3 229 8144037
BC105036.1 - 3 229 8144039
BC107882.1 - 3 197 6495107
BU520836.1 - 3 244
EU794626.1 - 3 263

View Full Product Details