Gene Transcripts

Gene Symbol
SLC12A1
Entrez Gene ID
6557
Gene Name
solute carrier family 12 member 1
Gene Aliases
BSC1, NKCC2
Chromosome Location
Chr. 15: 48206301 - 48304078 on Build GRCh38
UniGene ID
Hs.123116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000338.2 NP_000329.2 16 2192
NM_001184832.1 NP_001171761.1 16 2192
XM_005254606.1 XP_005254663.1 16 2192
GenBank mRNA AK295375.1 - 15 1645
AK298275.1 - 11 1411
AK298312.1 - 16 2191
BX647067.1 - 11 6115
BX647484.1 - 12 6099
EF559316.2 - 15 1990
U58130.1 - 15 1995

View Full Product Details