Gene Transcripts

Gene Symbol
SSR2
Entrez Gene ID
6746
Gene Name
signal sequence receptor subunit 2
Gene Aliases
HSD25, TLAP, TRAP-BETA, TRAPB
Chromosome Location
Chr. 1: 156009048 - 156020992 on Build GRCh38
UniGene ID
Hs.74564
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003145.3 NP_003136.1 6 912
XM_011509923.2 XP_011508225.1 6 937
GenBank mRNA AK092473.1 - 2 1622
AK222600.1 - 6 855
AK300678.1 - 6 912
AY251536.1 - 6 872
BC000341.2 - 6 858 2822983
D37991.1 - 6 856
X74104.1 - 6 882

View Full Product Details