Gene Transcripts

Gene Symbol
SLC4A8
Entrez Gene ID
9498
Gene Name
solute carrier family 4 member 8
Gene Aliases
NBC3, NDCBE
Chromosome Location
Chr. 12: 51391317 - 51515763 on Build GRCh38
UniGene ID
Hs.4749
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017020241.1 XP_016875730.1 22 3721
GenBank mRNA AB018282.1 - 24 3441
AF069512.1 - 22 3198
DQ996398.1 - 22 3411
DQ996537.1 - 22 3177

View Full Product Details