Gene Transcripts

Gene Symbol
MECOM
Entrez Gene ID
2122
Gene Name
MDS1 and EVI1 complex locus
Gene Aliases
AML1-EVI-1, EVI1, MDS1, MDS1-EVI1, PRDM3, RUSAT2
Chromosome Location
Chr. 3: 169083499 - 169663781 on Build GRCh38
UniGene ID
Hs.744090
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105077.3 NP_001098547.3 8 1318
NM_001105078.3 NP_001098548.2 7 1309
NM_001163999.1 NP_001157471.1 7 1256
NM_001164000.1 NP_001157472.1 7 2185
NM_001205194.1 NP_001192123.1 7 1613
NM_004991.3 NP_004982.2 8 1951
NM_005241.3 NP_005232.2 7 1253
XM_005247213.3 XP_005247270.1 8 1960
XM_005247214.3 XP_005247271.1 8 1960
XM_005247215.3 XP_005247272.1 8 1956
XM_005247219.2 XP_005247276.1 7 1925
XM_005247220.2 XP_005247277.1 7 1266
XM_005247221.2 XP_005247278.1 7 1249
XM_005247223.2 XP_005247280.1 7 1922
XM_011512546.2 XP_011510848.1 7 1713
XM_017005874.1 XP_016861363.1 7 2628
XM_017005875.1 XP_016861364.1 7 1263
XM_017005876.1 XP_016861365.1 7 1925
GenBank mRNA AK292865.1 - 7 1236
AK304098.1 - 8 1897
AK308404.1 - 8 1596
AK310509.1 - 6 1097
BC130520.1 - 8 1292 40146551
BC143951.1 - 8 1292 9052469
BC143952.1 - 7 1230 9052470
BX640908.1 - 7 2185
BX647613.1 - 7 1917
GQ352634.1 - 7 1517
X54989.1 - 7 1251

View Full Product Details