Gene Transcripts

Gene Symbol
PDIA3
Entrez Gene ID
2923
Gene Name
protein disulfide isomerase family A member 3
Gene Aliases
ER60, ERp57, ERp60, ERp61, GRP57, GRP58, HEL-S-269, HEL-S-93n, HsT17083, P58, PI-PLC
Chromosome Location
Chr. 15: 43746392 - 43772606 on Build GRCh38
UniGene ID
Hs.591095
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005313.4 NP_005304.3 9 1218
GenBank mRNA AK075455.1 - 10 1332
AK075481.1 - 10 1119
BC014433.1 - 9 1159 3542405
BC036000.1 - 9 1172 4712175
BC071878.1 - 9 1161 6380973
D83485.1 - 9 1092
EU668338.1 - 9 1172
FJ224330.1 - 9 1161
U42068.1 - 9 1148
Z49835.1 - 9 1176

Gene Symbol
PDIA3P1
Entrez Gene ID
171423
Gene Name
protein disulfide isomerase family A member 3 pseudogene 1
Gene Aliases
ERp60, GRP58P, PDIA3P
Chromosome Location
Chr. 1: 147177851 - 147179949 on Build GRCh38
UniGene ID
Hs.658566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002305.1 1 1333
GenBank mRNA AK295985.1 - 1 1218

View Full Product Details