Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190977.2 | NP_001177906.1 | 3 | 680 | |
NM_001190979.2 | NP_001177908.1 | 5 | 878 | ||
NM_001320080.1 | NP_001307009.1 | 5 | 906 | ||
NM_005748.5 | NP_005739.2 | 4 | 806 | ||
NR_135139.1 | 7 | 1048 | |||
NR_135140.1 | 7 | 1080 | |||
XM_006719184.3 | XP_006719247.1 | 4 | 725 | ||
XM_006719185.3 | XP_006719248.1 | 4 | 750 | ||
XM_011537728.2 | XP_011536030.1 | 6 | 905 | ||
XM_011537729.2 | XP_011536031.1 | 4 | 695 | ||
XM_017018669.1 | XP_016874158.1 | 6 | 864 | ||
GenBank mRNA | AK294260.1 | - | 3 | 601 | |
AK296191.1 | - | 5 | 786 | ||
BC037777.2 | - | 5 | 746 | 5276663 | |
BX649019.1 | - | 1 | 1115 |