Gene Transcripts

Gene Symbol
PAXIP1
Entrez Gene ID
22976
Gene Name
PAX interacting protein 1
Gene Aliases
CAGF28, CAGF29, PACIP1, PAXIP1L, PTIP, TNRC2
Chromosome Location
Chr. 7: 154943690 - 155003393 on Build GRCh38
UniGene ID
Hs.443881
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007349.3 NP_031375.3 6 906
XM_005249539.1 XP_005249596.1 7 1045
XM_011515982.2 XP_011514284.1 6 1020
XM_011515983.1 XP_011514285.1 6 906
XM_017011881.1 XP_016867370.1 7 2218
GenBank mRNA AK123600.1 - 7 1045
AK293744.1 - 4 738
AK307417.1 - 6 909
BC033781.1 - 1 147 4509264
BX640616.1 - 6 2804
U80735.1 - 1 261

View Full Product Details