Gene Transcripts

Gene Symbol
YBX2
Entrez Gene ID
51087
Gene Name
Y-box binding protein 2
Gene Aliases
CONTRIN, CSDA3, DBPC, MSY2
Chromosome Location
Chr. 17: 7288252 - 7294602 on Build GRCh38
UniGene ID
Hs.567494
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015982.3 NP_057066.2 9 1435
XM_017024713.1 XP_016880202.1 10 1525
GenBank mRNA AB208818.1 - 6 4416
AF096834.1 - 9 1433
AK092747.1 - 8 2052
BC033800.1 - 9 1398 5171696
BC047760.1 - 9 1399 5742121

View Full Product Details