Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304803.1 | NP_001291732.1 | 4 | 405 | |
NM_021947.2 | NP_068766.1 | 5 | 577 | ||
XM_006721565.3 | XP_006721628.1 | 5 | 609 | ||
XM_006721566.3 | XP_006721629.1 | 5 | 1787 | ||
XM_011523974.2 | XP_011522276.1 | 5 | 716 | ||
GenBank mRNA | AF169974.1 | - | 4 | 514 | |
AK023169.1 | - | 5 | 566 | ||
AK223120.1 | - | 5 | 570 | ||
AL834378.1 | - | 3 | 342 | ||
AY034081.1 | - | 4 | 498 | ||
BC074728.2 | - | 4 | 516 | 30915181 | |
CR457300.1 | - | 4 | 498 | ||
DA297969.1 | - | 4 | 394 |