Gene Transcripts

Gene Symbol
ALDH8A1
Entrez Gene ID
64577
Gene Name
aldehyde dehydrogenase 8 family member A1
Gene Aliases
ALDH12, DJ352A20.2
Chromosome Location
Chr. 6: 134917390 - 134950122 on Build GRCh38
UniGene ID
Hs.486520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193480.1 NP_001180409.1 6 2356
NM_022568.3 NP_072090.1 7 2506
NM_170771.2 NP_739577.1 6 2344
GenBank mRNA AF303134.1 - 7 2497
AK074266.1 - 6 2337
BX647324.1 - 1 2087

View Full Product Details