Gene Transcripts

Gene Symbol
JADE1
Entrez Gene ID
79960
Gene Name
jade family PHD finger 1
Gene Aliases
PHF17
Chromosome Location
Chr. 4: 128809623 - 128875224 on Build GRCh38
UniGene ID
Hs.12420
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287437.1 NP_001274366.1 6 781
NM_001287439.1 NP_001274368.1 6 935
NM_001287440.1 NP_001274369.1 6 732
NM_001287441.1 NP_001274370.1 6 696
NM_001287442.1 NP_001274371.1 6 689
NM_001287443.1 NP_001274372.1 6 693
NM_024900.4 NP_079176.2 6 817
NM_199320.3 NP_955352.1 6 817
XM_005263232.1 XP_005263289.1 6 659
XM_017008626.1 XP_016864115.1 6 774
XM_017008627.1 XP_016864116.1 6 821
GenBank mRNA AB058710.1 - 2 117
AF520952.1 - 6 714
AK027620.1 - 6 737
AK074986.1 - 1 44
AK127326.1 - 6 589
AK304235.1 - 6 650
AK304815.1 - 6 606
BC032376.1 - 6 651 5111727

View Full Product Details