Gene Transcripts

Gene Symbol
RDH12
Entrez Gene ID
145226
Gene Name
retinol dehydrogenase 12 (all-trans/9-cis/11-cis)
Gene Aliases
LCA13, RP53, SDR7C2
Chromosome Location
Chr. 14: 67701886 - 67734451 on Build GRCh38
UniGene ID
Hs.415322
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152443.2 NP_689656.2 9 1362
GenBank mRNA AK054835.1 - 7 1150
BC025724.1 - 7 1208 5199154

View Full Product Details