Gene Transcripts

Gene Symbol
TGM2
Entrez Gene ID
7052
Gene Name
transglutaminase 2
Gene Aliases
TG(C), TGC
Chromosome Location
Chr. 20: 38127387 - 38166578 on Build GRCh38
UniGene ID
Hs.517033
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323316.1 NP_001310245.1 10 1446
NM_001323317.1 NP_001310246.1 8 1220
NM_001323318.1 NP_001310247.1 8 1283
NM_004613.3 NP_004604.2 9 1463
NM_198951.2 NP_945189.1 9 1463
XM_011529028.1 XP_011527330.1 10 1408
GenBank mRNA AK058031.1 - 9 1231
AK291714.1 - 9 1304
AK295775.1 - 8 1061
AK300292.1 - 8 1124
AK314618.1 - 9 1304
AL512703.1 - 1 1006
AY675221.1 - 8 1232
BC003551.1 - 9 1316 3544757
BQ938957.1 - 1 24
EU794665.1 - 9 1332
M55153.1 - 9 1367
M98478.1 - 9 1364

View Full Product Details