Gene Transcripts
- Gene Symbol
- GPR89A
- Entrez Gene ID
- 653519
- Gene Name
- G protein-coupled receptor 89A
- Gene Aliases
- GPHR, GPR89, GPR89B, SH120, UNQ192
- Chromosome Location
- Chr. 1: 145607960 - 145670977 on Build GRCh38
- UniGene ID
- Hs.504320
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001097612.1 |
NP_001091081.1
|
4, 5 |
481 |
|
NM_001097613.2 |
NP_001091082.2
|
5, 6 |
527 |
|
XM_006711492.3 |
XP_006711555.1
|
4, 5 |
451 |
|
XM_011509908.2 |
XP_011508210.1
|
5, 6 |
497 |
|
XM_011509909.2 |
XP_011508211.1
|
4, 5 |
709 |
|
XM_011509910.2 |
XP_011508212.1
|
4, 5 |
451 |
|
XM_017002150.1 |
XP_016857639.1
|
4, 5 |
451 |
|
GenBank mRNA |
AB097024.1 |
- |
4, 5 |
422 |
|
AB097025.1 |
- |
4, 5 |
422 |
|
AF132947.1 |
- |
4, 5 |
406 |
|
AK223223.1 |
- |
4, 5 |
422 |
|
AK316371.1 |
- |
5, 6 |
439 |
|
AY358966.1 |
- |
4, 5 |
456 |
|
BC003187.1 |
- |
4, 5 |
384 |
3537949 |
BC146880.1 |
- |
4, 5 |
402 |
9056808 |
BT006679.1 |
- |
4, 5 |
307 |
|
KJ534847.1 |
- |
4, 5 |
307 |
|
KJ534848.1 |
- |
4, 5 |
307 |
|
KJ535029.1 |
- |
4, 5 |
307 |
|
- Gene Symbol
- GPR89B
- Entrez Gene ID
- 51463
- Gene Name
- G protein-coupled receptor 89B
- Gene Aliases
- GPHR, GPR89, GPR89C, SH120, UNQ192
- Chromosome Location
- Chr. 1: 147928393 - 147993523 on Build GRCh38
- UniGene ID
- Hs.504320
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016334.3 |
NP_057418.1
|
4, 5 |
451 |
|
XM_005277401.3 |
XP_005277458.1
|
4, 5 |
424 |
|
XM_005277402.4 |
XP_005277459.1
|
4, 5 |
588 |
|
XM_006711377.3 |
XP_006711440.1
|
5, 6 |
487 |
|
XM_011509610.2 |
XP_011507912.1
|
5, 6 |
484 |
|
XM_011509613.2 |
XP_011507915.1
|
4, 5 |
424 |
|
XM_017001448.1 |
XP_016856937.1
|
5, 6 |
987 |
|
XM_017001449.1 |
XP_016856938.1
|
4, 5 |
424 |
|
GenBank mRNA |
AK299995.1 |
- |
4, 5 |
422 |
|
AK302266.1 |
- |
4, 5 |
443 |
|
AL133020.1 |
- |
4, 5 |
435 |
|
DC305701.1 |
- |
4, 5 |
451 |
|
KJ534849.1 |
- |
4, 5 |
307 |
|
KJ534850.1 |
- |
4, 5 |
307 |
|
KJ535070.1 |
- |
5, 6 |
635 |
|
U78723.1 |
- |
4, 5 |
410 |
|
View Full Product Details