Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004581.5 | NP_004572.3 | 11, 12 | 1654 | |
NM_182836.2 | NP_878256.1 | 12, 13 | 1337 | ||
GenBank mRNA | AK291347.1 | - | 12, 13 | 1221 | |
AK292613.1 | - | 11, 12 | 1386 | ||
AK303247.1 | - | 9, 10 | 1269 | ||
AK308715.1 | - | 11, 12 | 1390 | ||
BC003093.1 | - | 12, 13 | 1275 | 3537388 | |
CR541797.1 | - | 10, 11 | 1142 | ||
Y08200.1 | - | 11, 12 | 1416 |