Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004581.5 | NP_004572.3 | 5 | 1035 | |
NM_182836.2 | NP_878256.1 | 6 | 718 | ||
GenBank mRNA | AK291347.1 | - | 6 | 602 | |
AK292613.1 | - | 5 | 767 | ||
AK303247.1 | - | 4 | 480 | ||
AK308715.1 | - | 6 | 601 | ||
AL547259.3 | - | 6 | 691 | ||
BC003093.1 | - | 6 | 656 | 3537388 | |
CR541797.1 | - | 4 | 523 | ||
Y08200.1 | - | 5 | 797 |