Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003940.2 | NP_003931.2 | 6 | 830 | |
XM_011513269.1 | XP_011511571.1 | 6 | 777 | ||
XM_017007425.1 | XP_016862914.1 | 6 | 674 | ||
XM_017007426.1 | XP_016862915.1 | 5 | 641 | ||
XM_017007427.1 | XP_016862916.1 | 3 | 361 | ||
GenBank mRNA | AK290338.1 | - | 6 | 777 | |
AK302404.1 | - | 6 | 639 | ||
BC016146.1 | - | 6 | 783 | 3910925 | |
HM138083.1 | - | 6 | 749 | ||
U75362.1 | - | 6 | 843 |