Gene Transcripts

Gene Symbol
PAMR1
Entrez Gene ID
25891
Gene Name
peptidase domain containing associated with muscle regeneration 1
Gene Aliases
DKFZP586H2123, FP938, RAMP
Chromosome Location
Chr. 11: 35431827 - 35530300 on Build GRCh38
UniGene ID
Hs.55044
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001991.2 NP_001001991.1 11 2529
NM_001282675.1 NP_001269604.1 13 2519
NM_001282676.1 NP_001269605.1 9 2196
NM_015430.3 NP_056245.2 12 2580
GenBank mRNA AF370388.1 - 10 2018
AK027841.1 - 12 2180
AK297092.1 - 13 2520
AK300185.1 - 9 1781
AK316188.1 - 9 1768
AL050214.1 - 1 467
AL832391.1 - 11 2095
AY358346.1 - 11 2108
BC089434.1 - 12 2172 30558617
BX640676.1 - 11 2529

View Full Product Details