Gene Transcripts

Gene Symbol
CPXM1
Entrez Gene ID
56265
Gene Name
carboxypeptidase X (M14 family), member 1
Gene Aliases
CPX1, CPXM
Chromosome Location
Chr. 20: 2794069 - 2800646 on Build GRCh38
UniGene ID
Hs.659346
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184699.1 NP_001171628.1 10 1371
NM_019609.4 NP_062555.1 10 1371
GenBank mRNA AK027661.1 - 10 1307
AK075508.1 - 6 583
AK075527.1 - 10 1346
AK303142.1 - 7 850
AK315027.1 - 10 1358
AY358956.1 - 10 1327
BC063430.1 - 10 1348 5239864

View Full Product Details