Gene Transcripts

Gene Symbol
TRPV5
Entrez Gene ID
56302
Gene Name
transient receptor potential cation channel subfamily V member 5
Gene Aliases
CAT2, ECAC1, OTRPC3
Chromosome Location
Chr. 7: 142908101 - 142933723 on Build GRCh38
UniGene ID
Hs.283369
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019841.6 NP_062815.3 15 2255
GenBank mRNA AF304464.1 - 15 2340
AJ271207.2 - 15 1991
AJ487965.2 - 15 1993

View Full Product Details