Gene Transcripts

Gene Symbol
NDUFV2
Entrez Gene ID
4729
Gene Name
NADH:ubiquinone oxidoreductase core subunit V2
Gene Aliases
CI-24k
Chromosome Location
Chr. 18: 9102623 - 9134345 on Build GRCh38
UniGene ID
Hs.464572
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021074.4 NP_066552.2 3, 4 291
XM_017025782.1 XP_016881271.1 3, 4 481
GenBank mRNA AK291865.1 - 3, 4 187
AK312129.1 - 3, 4 209
BC001632.1 - 3, 4 201 3536345
BC017487.1 - 3, 4 199 3537815
BC071689.1 - 3, 4 181 4372795
BG742948.1 - 3, 4 291
CR456928.1 - 3, 4 177
M22538.1 - 3, 4 195

View Full Product Details