Gene Transcripts

Gene Symbol
ERAP2
Entrez Gene ID
64167
Gene Name
endoplasmic reticulum aminopeptidase 2
Gene Aliases
L-RAP, LRAP
Chromosome Location
Chr. 5: 96875940 - 96919703 on Build GRCh38
UniGene ID
Hs.482910
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011543544.1 XP_011541846.1 2 736
GenBank mRNA AB109031.1 - 2 703
AB163917.1 - 2 703
AF191545.1 - 1 675
AK074281.1 - 2 780
AK312864.1 - 2 716
AY028805.1 - 1 633
BC017927.2 - 1 676
BC065240.1 - 2 697 6166289

View Full Product Details