Gene Transcripts

Gene Symbol
ERAP2
Entrez Gene ID
64167
Gene Name
endoplasmic reticulum aminopeptidase 2
Gene Aliases
L-RAP, LRAP
Chromosome Location
Chr. 5: 96875940 - 96919703 on Build GRCh38
UniGene ID
Hs.482910
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011543544.1 XP_011541846.1 5 1068
GenBank mRNA AB109031.1 - 5 1035
AB163917.1 - 5 1035
AF191545.1 - 4 1007
AK074281.1 - 5 1112
AK312864.1 - 5 1048
AY028805.1 - 4 965
BC017927.2 - 4 1008
BC065240.1 - 4 894 6166289

View Full Product Details