Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032802.3 | NP_116191.2 | 2 | 270 | |
XM_005254722.3 | XP_005254779.1 | 2 | 269 | ||
XM_017022680.1 | XP_016878169.1 | 2 | 269 | ||
XM_017022681.1 | XP_016878170.1 | 2 | 269 | ||
GenBank mRNA | AJ345028.1 | - | 2 | 248 | |
AK075454.1 | - | 2 | 259 | ||
AK315651.1 | - | 2 | 237 | ||
AY169314.1 | - | 2 | 266 | ||
BC025740.1 | - | 2 | 233 | 5223715 | |
DA023801.1 | - | 2 | 270 |