Gene Transcripts

Gene Symbol
NEO1
Entrez Gene ID
4756
Gene Name
neogenin 1
Gene Aliases
IGDCC2, NGN, NTN1R2
Chromosome Location
Chr. 15: 73051715 - 73305206 on Build GRCh38
UniGene ID
Hs.388613
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172623.1 NP_001166094.1 6 1259
NM_001172624.1 NP_001166095.1 6 1259
NM_002499.3 NP_002490.2 6 1259
XM_005254408.1 XP_005254465.1 7 1509
XM_011521628.2 XP_011519930.1 6 1264
XM_011521629.2 XP_011519931.1 6 1258
XM_011521630.2 XP_011519932.1 6 1258
XM_011521632.2 XP_011519934.1 6 1258
XM_011521633.2 XP_011519935.1 6 1258
XM_017022231.1 XP_016877720.1 6 1258
XM_017022232.1 XP_016877721.1 7 1509
XM_017022233.1 XP_016877722.1 6 1258
XM_017022234.1 XP_016877723.1 7 1301
XM_017022235.1 XP_016877724.1 6 1258
XM_017022236.1 XP_016877725.1 6 1258
XM_017022237.1 XP_016877726.1 6 1258
XM_017022238.1 XP_016877727.1 6 1258
GenBank mRNA AB209412.1 - 2 95
BC117161.1 - 6 1133 40125712
BC143270.1 - 6 1134 9051772
BC143271.1 - 6 1133 9051773
U61262.1 - 6 1203
U72391.1 - 6 1193

View Full Product Details