Gene Transcripts

Gene Symbol
PTPRZ1
Entrez Gene ID
5803
Gene Name
protein tyrosine phosphatase, receptor type Z1
Gene Aliases
HPTPZ, HPTPzeta, PTP-ZETA, PTP18, PTPRZ, PTPZ, R-PTP-zeta-2, RPTPB, RPTPbeta, phosphacan
Chromosome Location
Chr. 7: 121873105 - 122062036 on Build GRCh38
UniGene ID
Hs.489824
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002851.2 NP_002842.2 12 4346
XM_005250519.1 XP_005250576.1 12 4346
XM_017012477.1 XP_016867966.1 13 4393
GenBank mRNA M93426.1 - 12 4098

View Full Product Details