Gene Transcripts

Gene Symbol
POU4F1
Entrez Gene ID
5457
Gene Name
POU class 4 homeobox 1
Gene Aliases
BRN3A, Oct-T1, RDC-1, brn-3A
Chromosome Location
Chr. 13: 78599092 - 78603560 on Build GRCh38
UniGene ID
Hs.654522
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006237.3 NP_006228.3 2 2432
GenBank mRNA L20433.1 - 2 2435
X64624.1 - 1 2203

View Full Product Details