Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007240.2 | NP_009171.1 | 5 | 856 | |
XM_005244862.2 | XP_005244919.1 | 5 | 945 | ||
GenBank mRNA | AF119226.1 | - | 5 | 833 | |
AK000259.1 | - | 3 | 270 | ||
BC006286.1 | - | 5 | 854 | 3958403 | |
BT006633.1 | - | 5 | 822 |