Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001025247.1 | NP_001020418.1 | 4 | 435 | |
NM_014409.3 | NP_055224.1 | 4 | 435 | ||
XM_005273099.4 | XP_005273156.1 | 4 | 388 | ||
XM_005273100.2 | XP_005273157.1 | 5 | 401 | ||
XM_011544162.1 | XP_011542464.1 | 3 | 172 | ||
XM_011544163.1 | XP_011542465.1 | 7 | 1017 | ||
XM_011544164.2 | XP_011542466.1 | 3 | 115 | ||
XM_017001045.1 | XP_016856534.1 | 6 | 1172 | ||
GenBank mRNA | AF069736.1 | - | 3 | 269 | |
BC041094.1 | - | 4 | 358 | 5551246 |