Gene Transcripts

Gene Symbol
ARNTL2
Entrez Gene ID
56938
Gene Name
aryl hydrocarbon receptor nuclear translocator like 2
Gene Aliases
BMAL2, CLIF, MOP9, PASD9, bHLHe6
Chromosome Location
Chr. 12: 27332854 - 27425813 on Build GRCh38
UniGene ID
Hs.445447
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001248002.1 NP_001234931.1 9 1287
NM_001248003.1 NP_001234932.1 8 1218
NM_001248004.1 NP_001234933.1 8 1185
NM_001248005.1 NP_001234934.1 8 1218
NM_020183.4 NP_064568.3 10 1329
XM_006719112.3 XP_006719175.1 9 1288
XM_006719114.3 XP_006719177.1 9 1019
XM_011520766.2 XP_011519068.1 10 1330
XM_011520768.2 XP_011519070.1 9 1238
XM_011520769.2 XP_011519071.1 9 1228
XM_011520770.2 XP_011519072.1 10 1148
XM_011520771.2 XP_011519073.1 10 1114
XM_017019666.1 XP_016875155.1 7 1059
XM_017019667.1 XP_016875156.1 10 1346
XM_017019668.1 XP_016875157.1 10 1143
XM_017019669.1 XP_016875158.1 9 990
GenBank mRNA AB039921.1 - 7 861
AF231338.1 - 8 1014
AF231339.1 - 7 975
AF246960.1 - 10 1128
AF246961.1 - 9 1086
AF246962.1 - 8 1017
AF246963.1 - 8 984
AF256215.1 - 1227
AK296706.1 - 8 1152
BC000172.3 - 7 936
BC125061.1 - 8 979 40117267
BC125062.1 - 8 1017 40117268

View Full Product Details