Gene Transcripts

Gene Symbol
RFX5
Entrez Gene ID
5993
Gene Name
regulatory factor X5
Gene Aliases
Chromosome Location
Chr. 1: 151340640 - 151347319 on Build GRCh38
UniGene ID
Hs.632472
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000449.3 NP_000440.1 11 2402
NM_001025603.1 NP_001020774.1 11 2395
XM_005245405.1 XP_005245462.1 11 2457
XM_005245406.3 XP_005245463.1 11 2414
XM_011509847.1 XP_011508149.1 11 2461
XM_011509848.1 XP_011508150.1 11 2464
XM_011509849.1 XP_011508151.1 11 2397
XM_011509850.1 XP_011508152.1 11 2524
XM_017001999.1 XP_016857488.1 11 2525
XM_017002000.1 XP_016857489.1 11 2556
GenBank mRNA AL050135.1 - 3 1637

View Full Product Details