Gene Transcripts

Gene Symbol
KLF9
Entrez Gene ID
687
Gene Name
Kruppel like factor 9
Gene Aliases
BTEB, BTEB1
Chromosome Location
Chr. 9: 70384597 - 70414657 on Build GRCh38
UniGene ID
Hs.150557
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206.2 NP_001197.1 2 2753
GenBank mRNA D31716.1 - 2 2720

View Full Product Details