Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005171.4 | NP_005162.1 | 7 | 1251 | |
XM_011538386.2 | XP_011536688.1 | 7 | 3308 | ||
XM_011538387.2 | XP_011536689.1 | 7 | 981 | ||
XM_011538388.2 | XP_011536690.1 | 7 | 1182 | ||
XM_017019331.1 | XP_016874820.1 | 7 | 1021 | ||
XM_017019332.1 | XP_016874821.1 | 7 | 1115 | ||
XM_017019333.1 | XP_016874822.1 | 7 | 1001 | ||
XM_017019334.1 | XP_016874823.1 | 7 | 1027 | ||
XM_017019335.1 | XP_016874824.1 | 7 | 1025 | ||
XM_017019336.1 | XP_016874825.1 | 7 | 1020 | ||
GenBank mRNA | AK299240.1 | - | 6 | 867 | |
BC029619.1 | - | 7 | 965 | 4102231 | |
BG611151.1 | - | 3 | 467 | ||
BG611861.1 | - | 1 | 180 | ||
BX102171.1 | - | 3 | 487 |