Gene Transcripts

Gene Symbol
STAT2
Entrez Gene ID
6773
Gene Name
signal transducer and activator of transcription 2
Gene Aliases
IMD44, ISGF-3, P113, STAT113
Chromosome Location
Chr. 12: 56341597 - 56360253 on Build GRCh38
UniGene ID
Hs.530595
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005419.3 NP_005410.1 11, 12 1305
NM_198332.1 NP_938146.1 11, 12 1293
XM_005269111.3 XP_005269168.2 11, 12 1213
XM_011538697.2 XP_011536999.1 11, 12 1213
XM_011538698.2 XP_011537000.1 11, 12 1174
XM_011538699.2 XP_011537001.1 11, 12 1213
XM_011538700.2 XP_011537002.1 5, 6 611
XM_017019904.1 XP_016875393.1 10, 11 1672
GenBank mRNA AK296939.1 - 11, 12 1166
AK296940.1 - 11, 12 1166
BC006092.1 - 10, 11 1092
BC051284.1 - 10, 11 1125 6460486
M97934.1 - 11, 12 1139

View Full Product Details