Gene Transcripts

Gene Symbol
NFRKB
Entrez Gene ID
4798
Gene Name
nuclear factor related to kappaB binding protein
Gene Aliases
INO80G
Chromosome Location
Chr. 11: 129863773 - 129895596 on Build GRCh38
UniGene ID
Hs.530539
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143835.1 NP_001137307.1 8 977
NM_006165.3 NP_006156.2 6 972
XM_005271567.3 XP_005271624.1 8 1013
XM_005271568.3 XP_005271625.1 7 928
XM_011542850.2 XP_011541152.1 9 1050
XM_011542851.2 XP_011541153.1 8 1032
XM_011542852.2 XP_011541154.1 7 870
XM_017017793.1 XP_016873282.1 7 924
XM_017017794.1 XP_016873283.1 7 888
XM_017017795.1 XP_016873284.1 9 1014
XM_017017796.1 XP_016873285.1 7 892
XM_017017797.1 XP_016873286.1 7 924
XM_017017798.1 XP_016873287.1 8 977
XM_017017799.1 XP_016873288.1 9 1124
XM_017017800.1 XP_016873289.1 9 1064
GenBank mRNA BC063280.1 - 8 923
X80878.1 - 6 981

View Full Product Details