Gene Transcripts

Gene Symbol
ZNF75D
Entrez Gene ID
7626
Gene Name
zinc finger protein 75D
Gene Aliases
D8C6, ZKSCAN24, ZNF75, ZNF82, ZSCAN28
Chromosome Location
Chr. X: 135248589 - 135344109 on Build GRCh38
UniGene ID
Hs.371717
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001185063.2 NP_001171992.1 4 1992
NM_007131.4 NP_009062.2 7 4597
XM_005262469.1 XP_005262526.1 7 3126
XM_005262471.3 XP_005262528.1 5 3572
XM_011531391.1 XP_011529693.1 8 3313
XM_011531395.2 XP_011529697.1 4 3726
XM_011531396.2 XP_011529698.1 6 4090
XM_011531397.2 XP_011529699.1 5 3998
XM_017029818.1 XP_016885307.1 6 4527
XM_017029819.1 XP_016885308.1 4 7292
XM_017029820.1 XP_016885309.1 5 2505
GenBank mRNA AK091687.1 - 4 1986
AK092347.1 - 4 1486
AK290215.1 - 6 2301
AL831984.2 - 7 4542
AL832801.1 - 6 2084
BC109100.2 - 6 2116 40034784

View Full Product Details