Gene Transcripts

Gene Symbol
GYG1
Entrez Gene ID
2992
Gene Name
glycogenin 1
Gene Aliases
GSD15, GYG
Chromosome Location
Chr. 3: 148991408 - 149027669 on Build GRCh38
UniGene ID
Hs.477892
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184720.1 NP_001171649.1 4, 5 706
NM_001184721.1 NP_001171650.1 4, 5 706
NM_004130.3 NP_004121.2 4, 5 706
XM_017006275.1 XP_016861764.1 3, 4 463
GenBank mRNA AF087942.1 - 4, 5 526
AK312283.1 - 4, 5 542
BC000033.2 - 4, 5 562 3504538
BC031096.2 - 3, 4 467
CR536547.1 - 3, 4 473
U31525.1 - 4, 5 600
U44131.1 - 3, 4 481
X79537.1 - 4, 5 556

View Full Product Details