Gene Transcripts
- Gene Symbol
- SLC35A3
- Entrez Gene ID
- 23443
- Gene Name
- solute carrier family 35 member A3
- Gene Aliases
- AMRS
- Chromosome Location
- Chr. 1: 99969367 - 100026979 on Build GRCh38
- UniGene ID
- Hs.448979
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271684.1 |
NP_001258613.1
|
3, 4 |
731 |
|
NM_001271685.1 |
NP_001258614.1
|
3, 4 |
566 |
|
NM_012243.2 |
NP_036375.1
|
3, 4 |
731 |
|
XM_005270691.4 |
XP_005270748.1
|
4, 5 |
1252 |
|
XM_011541136.2 |
XP_011539438.1
|
4, 5 |
648 |
|
XM_011541137.2 |
XP_011539439.1
|
3, 4 |
708 |
|
XM_011541138.2 |
XP_011539440.1
|
3, 4 |
717 |
|
XM_017000870.1 |
XP_016856359.1
|
3, 4 |
1153 |
|
GenBank mRNA |
AB021981.1 |
- |
3, 4 |
536 |
|
AK024989.1 |
- |
2, 3 |
296 |
|
AK290573.1 |
- |
3, 4 |
489 |
|
BC005136.2 |
- |
3, 4 |
514 |
3505772 |
CR749816.1 |
- |
3, 4 |
566 |
|
View Full Product Details