Gene Transcripts

Gene Symbol
PABPC1
Entrez Gene ID
26986
Gene Name
poly(A) binding protein cytoplasmic 1
Gene Aliases
PAB1, PABP, PABP1, PABPC2, PABPL1
Chromosome Location
Chr. 8: 100702916 - 100722998 on Build GRCh38
UniGene ID
Hs.387804
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002568.3 NP_002559.2 15 2433
XM_005250861.2 XP_005250918.1 15 3361
GenBank mRNA AK095194.1 - 15 2268
AK130523.1 - 15 2433
AK298990.1 - 16 2149
AK303120.1 - 16 2157
BC015958.2 - 16 2328 3940309
BC023520.2 - 15 2412 4123269
BC041863.1 - 15 3057
Y00345.1 - 15 2422

View Full Product Details