Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004521.2 | NP_004512.1 | 2 | 610 | |
XM_017016224.1 | XP_016871713.1 | 2 | 216 | ||
GenBank mRNA | AK289413.1 | - | 2 | 556 | |
AK307502.1 | - | 3 | 307 | ||
BC010885.1 | - | 2 | 546 | ||
BC065267.1 | - | 2 | 567 | ||
BC080604.1 | - | 2 | 541 | ||
BC111042.1 | - | 2 | 550 | ||
BC126279.1 | - | 2 | 363 | 8991995 | |
BC126281.1 | - | 2 | 363 | 8991997 | |
FJ224324.1 | - | 2 | 453 | ||
X65873.1 | - | 2 | 453 |